LIF945291637 Life Chemicals Inc.

[1-(3-fluoro-4-methylphenyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD16747678
  • Cas Number : 1250228-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart