LIF945291639 Life Chemicals Inc.

[1-(4-fluoro-2-methylphenyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD16747688
  • Cas Number : 1247731-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart