LIF945291640 Life Chemicals Inc.

[1-(2-fluoro-4-methylphenyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD16747696
  • Cas Number : 1096130-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart