LIF945291645 Life Chemicals Inc.

{1-[2-(methylsulfanyl)phenyl]-1H-1,2,3-triazol-4-yl}methanol

  • MDL number MFCD16747694
  • Cas Number : 1249284-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart