LIF945291664 Life Chemicals Inc.

methyl 2,2-dimethylbutanoate

  • MDL number MFCD16844899
  • Cas Number : 813-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart