LIF945291666 Life Chemicals Inc.

2-amino-1-(2-chloro-6-fluorophenyl)ethan-1-one hydrochloride

  • MDL number MFCD22665968
  • Cas Number : 1909317-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart