LIF945291668 Life Chemicals Inc.

2-amino-1-(2,5-dichlorophenyl)ethan-1-one hydrochloride

  • MDL number MFCD06739762
  • Cas Number : 51084-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart