LIF945291673 Life Chemicals Inc.

3-methyl-4-(methylamino)phenol hydrochloride

  • MDL number MFCD30486206
  • Cas Number : 2034156-77-7
  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart