LIF945291675 Life Chemicals Inc.

[2-(4-fluorophenyl)-4-methyl-1,3-thiazol-5-yl]methanol

  • MDL number MFCD11937737
  • Cas Number : 317319-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart