LIF945291676 Life Chemicals Inc.

3-hydroxy-2-methoxybenzaldehyde

  • MDL number MFCD06656119
  • Cas Number : 66495-88-3
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart