LIF945291679 Life Chemicals Inc.

2-(3-aminopropoxy)ethan-1-ol

  • MDL number MFCD11618156
  • Cas Number : 99513-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart