LIF945291685 Life Chemicals Inc.

5-(2,5-dichlorophenyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07376207
  • Cas Number : 1249213-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart