LIF945291698 Life Chemicals Inc.

4-(4-methylphenoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD01631869
  • Cas Number : 192329-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart