LIF945291702 Life Chemicals Inc.

2-(1-methyl-1H-imidazol-2-yl)piperidine

  • MDL number MFCD10700194
  • Cas Number : 933713-92-9
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart