LIF945291703 Life Chemicals Inc.

4-(oxolan-3-yl)-1H-pyrazole

  • MDL number MFCD30495015
  • Cas Number : 2097937-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart