LIF945291706 Life Chemicals Inc.

1-[(benzylamino)methyl]cyclohexan-1-ol

  • MDL number MFCD12811676
  • Cas Number : 113912-41-7
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart