LIF945291711 Life Chemicals Inc.

(2S)-2-amino-5-(N-hydroxyamino)pentanoic acid hydrochloride

  • MDL number MFCD30496505
  • Cas Number : 37552-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart