LIF945291717 Life Chemicals Inc.

2-(2,3-dimethoxyphenyl)acetic acid

  • MDL number MFCD00060351
  • Cas Number : 90-53-9
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart