LIF945291722 Life Chemicals Inc.

3-(4-methylphenoxy)benzoic acid

  • MDL number MFCD03840106
  • Cas Number : 62507-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart