LIF945291725 Life Chemicals Inc.

4-methoxy-4-(trifluoromethyl)piperidine hydrochloride

  • MDL number MFCD29060459
  • Cas Number : 1332886-70-0
  • Molecular Weight : 219.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart