LIF945291741 Life Chemicals Inc.

2-amino-3,5-dibromopyridin-4-ol

  • MDL number MFCD18253542
  • Cas Number : 1261269-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart