LIF945291747 Life Chemicals Inc.

sodium 2-oxaspiro[3.3]heptane-6-carboxylate

  • MDL number MFCD30493354
  • Cas Number : 2095409-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart