LIF945291749 Life Chemicals Inc.

tert-butyl 6-methylpiperidine-3-carboxylate

  • MDL number MFCD29060431
  • Cas Number : 1909326-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart