LIF945291750 Life Chemicals Inc.

tert-butyl 4-methylpiperidine-3-carboxylate

  • MDL number MFCD30180350
  • Cas Number : 1936339-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart