LIF945291753 Life Chemicals Inc.

2-(furan-2-yl)benzaldehyde

  • MDL number MFCD03990475
  • Cas Number : 16191-32-5
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart