LIF945291754 Life Chemicals Inc.

2-[(difluoromethyl)sulfanyl]aniline

  • MDL number MFCD06655213
  • Cas Number : 24933-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart