LIF945291755 Life Chemicals Inc.

[1-methyl-2-(methylsulfanyl)-1H-imidazol-5-yl]methanol

  • MDL number MFCD02179533
  • Cas Number : 107718-01-4
  • Molecular Weight : 158.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart