LIF945291758 Life Chemicals Inc.

1-[(1H-pyrazol-1-yl)methyl]cyclohexan-1-ol

  • MDL number MFCD21182486
  • Cas Number : 1496253-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart