LIF945291759 Life Chemicals Inc.

1-[(1H-pyrazol-1-yl)methyl]cyclopentan-1-ol

  • MDL number MFCD21182632
  • Cas Number : 1464933-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart