LIF945291774 Life Chemicals Inc.

methyl 2-oxoacetate hydrate

  • MDL number MFCD27578368
  • Cas Number : 1401921-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart