LIF945291782 Life Chemicals Inc.

3-benzylpyrrolidine hydrochloride

  • MDL number MFCD08460458
  • Cas Number : 936225-49-9
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart