LIF945291783 Life Chemicals Inc.

(2S)-2-amino-3-(1H-1,2,3-benzotriazol-1-yl)propanoic acid hydrochloride

  • MDL number MFCD09971775
  • Cas Number : 1212326-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart