LIF945291784 Life Chemicals Inc.

5-ethoxy-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD08273570
  • Cas Number : 885371-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart