LIF945291791 Life Chemicals Inc.

1-amino-3-fluoropropan-2-ol hydrochloride

  • MDL number MFCD28975644
  • Cas Number : 53460-74-5
  • Molecular Weight : 129.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart