LIF945291793 Life Chemicals Inc.

5-{imidazo[1,2-a]pyrimidin-2-yl}-2-methoxyaniline

  • MDL number MFCD07403535
  • Cas Number : 879069-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart