LIF945291801 Life Chemicals Inc.

4-(2-methyl-1H-imidazol-1-yl)-6-(piperazin-1-yl)pyrimidine

  • MDL number MFCD28012627
  • Cas Number : 1706446-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart