LIF945291802 Life Chemicals Inc.

2-methyl-4-(piperazin-1-yl)-6-(pyrrolidin-1-yl)pyrimidine

  • MDL number MFCD28012651
  • Cas Number : 1706447-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart