LIF945291807 Life Chemicals Inc.

2-cyclopropyl-1H-imidazole

  • MDL number MFCD19217297
  • Cas Number : 89532-38-7
  • Molecular Weight : 108.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart