LIF945291813 Life Chemicals Inc.

2-(furan-3-yl)ethan-1-amine

  • MDL number MFCD08448832
  • Cas Number : 53916-75-9
  • Molecular Weight : 111.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart