LIF945291815 Life Chemicals Inc.

5-methoxy-3-methylpyridin-2-amine

  • MDL number MFCD18250734
  • Cas Number : 1101060-84-7
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart