LIF945291820 Life Chemicals Inc.

2-(chloromethyl)-1,3,4-oxadiazole

  • MDL number MFCD01109233
  • Cas Number : 73315-63-6
  • Molecular Weight : 118.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart