LIF945291851 Life Chemicals Inc.

4-(3-methylphenyl)butanoic acid

  • MDL number MFCD06810497
  • Cas Number : 22156-45-2
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart