LIF945291852 Life Chemicals Inc.

4-(3-methylphenyl)-4-oxobutanoic acid

  • MDL number MFCD01320043
  • Cas Number : 59618-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart