LIF945291853 Life Chemicals Inc.

2,3-dihydro-1-benzofuran-3-carboxamide

  • MDL number MFCD19655494
  • Cas Number : 1343275-28-4
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart