LIF945291854 Life Chemicals Inc.

5-methyl-3-(thiophen-2-yl)-1H-pyrazole

  • MDL number MFCD00514532
  • Cas Number : 1224742-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart