LIF945291863 Life Chemicals Inc.

4-[(2-formyl-3-hydroxyphenoxy)methyl]benzoic acid

  • MDL number MFCD00868063
  • Cas Number : 84290-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart