LIF945291874 Life Chemicals Inc.

2-oxo-2H-pyran-6-carboxylic acid

  • MDL number MFCD00177095
  • Cas Number : 672-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart