LIF945291878 Life Chemicals Inc.

4,4-dimethyl-1,2,3,4-tetrahydroisoquinolin-1-one

  • MDL number MFCD19347428
  • Cas Number : 27295-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart