LIF945291879 Life Chemicals Inc.

3-bromo-1,5-naphthyridine

  • MDL number MFCD11858569
  • Cas Number : 17965-71-8
  • Molecular Weight : 209.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart