LIF945291881 Life Chemicals Inc.

N,6-dimethyl-N-(piperidin-4-yl)pyrimidin-4-amine

  • MDL number MFCD30738808
  • Cas Number : 1553946-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart